ENA018105274 Enamine

trifluoroethanecarbonyl isocyanate

  • MDL number MFCD01320688
  • Cas Number : 14565-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-01256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01256
Availability details for
Quantity
  • Availability
  • Add to Cart