ENA018105275 Enamine

2-Isocyanato-benzonitrile

  • MDL number MFCD00829130
  • Cas Number : 42066-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01257

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01257
Availability details for
Quantity
  • Availability
  • Add to Cart