ENA018105276 Enamine

2-chloro-1-(difluoromethoxy)-4-isothiocyanatobenzene

  • MDL number MFCD06660584
  • Cas Number : 865149-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01258

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01258
Availability details for
Quantity
  • Availability
  • Add to Cart