ENA018105322 Enamine

2-Cyano-N-(2-methyl-cyclohexyl)-acetamide

  • MDL number MFCD02720725
  • Cas Number : 380190-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01312

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01312
Availability details for
Quantity
  • Availability
  • Add to Cart