ENA018105337 Enamine

2-ethyl-3-methylquinoline-4-carboxylic acid

  • MDL number MFCD02231996
  • Cas Number : 74960-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 287-288
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01328
Availability details for
Quantity
  • Availability
  • Add to Cart