ENA018105345 Enamine

2-benzyl-1H,2H,3H,4H-benzo[b]1,6-naphthyridine-10-carboxylic acid

  • MDL number MFCD01911471
  • Cas Number : 99117-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260-261
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01338
Availability details for
Quantity
  • Availability
  • Add to Cart