ENA018105361 Enamine

N,N'-Dicyclohexyl-malonamide

  • MDL number MFCD00431335
  • Cas Number : 10256-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01354
Availability details for
Quantity
  • Availability
  • Add to Cart