ENA018105362 Enamine

N,N'-bis(2-phenylethyl)propanediamide

  • MDL number MFCD01060446
  • Cas Number : 67942-22-7
  • Molecular Weight : 310.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-01355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01355
Availability details for
Quantity
  • Availability
  • Add to Cart