ENA018105368 Enamine

2-Cyano-N-(tetrahydro-furan-2-ylmethyl)-acetamide

  • MDL number MFCD00121391
  • Cas Number : 324546-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91...93
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01361

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01361
Availability details for
Quantity
  • Availability
  • Add to Cart