ENA018105373 Enamine

N-{5-[(2-chlorophenyl)methyl]-1,3-thiazol-2-yl}-2-cyanoacetamide

  • MDL number MFCD01918088
  • Cas Number : 307514-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-01367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01367
Availability details for
Quantity
  • Availability
  • Add to Cart