ENA018105391 Enamine

2-(benzylamino)-4,5-dihydro-1,3-thiazol-4-one

  • MDL number MFCD03152772
  • Cas Number : 17385-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01388
Availability details for
Quantity
  • Availability
  • Add to Cart