ENA018105414 Enamine

4-(4-Bromo-phenyl)-2-methyl-thiazole

  • MDL number MFCD00098207
  • Cas Number : 66047-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01411

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01411
Availability details for
Quantity
  • Availability
  • Add to Cart