ENA018105418 Enamine

2-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}acetonitrile

  • MDL number MFCD00980781
  • Cas Number : 116598-69-7
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-66
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01415
Availability details for
Quantity
  • Availability
  • Add to Cart