ENA018105436 Enamine

N,N'-Bis-(4-fluoro-phenyl)-malonamide

  • MDL number MFCD00118891
  • Cas Number : 1677-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217...219
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01433

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01433
Availability details for
Quantity
  • Availability
  • Add to Cart