ENA018105497 Enamine

4-ethyl 2-methyl 5-(2-chloroacetamido)-3-methylthiophene-2,4-dicarboxylate

  • MDL number MFCD01911691
  • Cas Number : 300676-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-01500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01500
Availability details for
Quantity
  • Availability
  • Add to Cart