ENA018105524 Enamine

4-Chloro-N-(4-methoxy-phenyl)-3-nitro-benzenesulfonamide

  • MDL number MFCD01269337
  • Cas Number : 30751-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01556

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01556
Availability details for
Quantity
  • Availability
  • Add to Cart