ENA018105525 Enamine

2-Chloro-N,N-diethyl-5-nitro-benzenesulfonamide

  • MDL number MFCD02724572
  • Cas Number : 4750-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01557

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01557
Availability details for
Quantity
  • Availability
  • Add to Cart