ENA018105529 Enamine

5-tert-Butyl-2-chloromethyl-1,3-dimethyl-benzene

  • MDL number MFCD03147319
  • Cas Number : 19387-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01563
Availability details for
Quantity
  • Availability
  • Add to Cart