ENA018105538 Enamine

2-Chloro-N-(2-chloro-5-diethylsulfamoyl-phenyl)-acetamide

  • MDL number MFCD02698355
  • Cas Number : 325764-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01611

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01611
Availability details for
Quantity
  • Availability
  • Add to Cart