ENA018105544 Enamine

2-Chloro-N-[2-methyl-5-(morpholine-4-sulfonyl)-phenyl]-acetamide

  • MDL number MFCD03147353
  • Cas Number : 379255-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01628

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01628
Availability details for
Quantity
  • Availability
  • Add to Cart