ENA018105545 Enamine

2-chloro-N-[4-(morpholine-4-sulfonyl)phenyl]acetamide

  • MDL number MFCD02580955
  • Cas Number : 35959-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-01633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01633
Availability details for
Quantity
  • Availability
  • Add to Cart