ENA018105561 Enamine

2-chloro-1-[5-(4-methoxyphenyl)-3-phenyl-4,5-dihydro-1H-pyrazol-1-yl]ethan-1-one

  • MDL number MFCD02720609
  • Cas Number : 380196-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-01707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01707
Availability details for
Quantity
  • Availability
  • Add to Cart