ENA018105572 Enamine

2,4-dichloro-5-(chlorosulfonyl)benzoic acid

  • MDL number MFCD00486543
  • Cas Number : 3740-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-185
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-01720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01720
Availability details for
Quantity
  • Availability
  • Add to Cart