ENA018105582 Enamine

2-Chloro-N-(4-chloro-phenyl)-5-nitro-benzenesulfonamide

  • MDL number MFCD00682816
  • Cas Number : 380342-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01738

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01738
Availability details for
Quantity
  • Availability
  • Add to Cart