ENA018105589 Enamine

5-tert-butyl-2-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD03147400
  • Cas Number : 63452-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01796
Availability details for
Quantity
  • Availability
  • Add to Cart