ENA018105606 Enamine

2,6-dichloro-3-(chlorosulfonyl)benzoic acid

  • MDL number MFCD06660592
  • Cas Number : 53553-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01839
Availability details for
Quantity
  • Availability
  • Add to Cart