ENA018105607 Enamine

3-(chlorosulfonyl)-2,6-difluorobenzoic acid

  • MDL number MFCD03030334
  • Cas Number : 142576-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01840
Availability details for
Quantity
  • Availability
  • Add to Cart