ENA018105612 Enamine

3-Chlorosulfonyl-4,5-dimethyl-benzoic acid

  • MDL number MFCD06654899
  • Cas Number : 151104-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01847

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01847
Availability details for
Quantity
  • Availability
  • Add to Cart