ENA018105621 Enamine

2-Chloro-N-[5-(morpholine-4-sulfonyl)-2-morpholin-4-yl-phenyl]-acetamide

  • MDL number MFCD03955641
  • Cas Number : 721908-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01860

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01860
Availability details for
Quantity
  • Availability
  • Add to Cart