ENA018105627 Enamine

methyl 2-(2-chloroacetamido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD00610983
  • Cas Number : 300559-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-01869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01869
Availability details for
Quantity
  • Availability
  • Add to Cart