ENA018105648 Enamine

2-chloro-1-(9H-fluoren-3-yl)ethan-1-one

  • MDL number MFCD06362442
  • Cas Number : 792953-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-134
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-01892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01892
Availability details for
Quantity
  • Availability
  • Add to Cart