ENA018105655 Enamine

2-Chloro-N-(4'-cyano-[2,2';3',2'']terfuran-5'-yl)-acetamide

  • MDL number MFCD03965251
  • Cas Number : 554404-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01901

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01901
Availability details for
Quantity
  • Availability
  • Add to Cart