ENA018105664 Enamine

3-(2-Bromo-acetyl)-8-methoxy-chromen-2-one

  • MDL number MFCD01246894
  • Cas Number : 106578-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01917

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01917
Availability details for
Quantity
  • Availability
  • Add to Cart