ENA018105672 Enamine

2-chloro-N-(3-cyano-6-methyl-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)acetamide

  • MDL number MFCD01067832
  • Cas Number : 331869-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-01933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01933
Availability details for
Quantity
  • Availability
  • Add to Cart