ENA018105697 Enamine

2-chloro-N-(5-phenyl-1,3,4-oxadiazol-2-yl)acetamide

  • MDL number MFCD01710644
  • Cas Number : 7659-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01974
Availability details for
Quantity
  • Availability
  • Add to Cart