ENA018105701 Enamine

2-Chloro-N-[4-(4-ethyl-phenyl)-thiazol-2-yl]-propionamide

  • MDL number MFCD03966878
  • Cas Number : 728908-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01978

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01978
Availability details for
Quantity
  • Availability
  • Add to Cart