ENA018105706 Enamine

1-(4-sec-Butyl-phenyl)-2-chloro-ethanone

  • MDL number MFCD03966850
  • Cas Number : 77234-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01983

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01983
Availability details for
Quantity
  • Availability
  • Add to Cart