ENA018105717 Enamine

4-Chloro-N-(3-chloro-quinoxalin-2-yl)-benzenesulfonamide

  • MDL number MFCD03651821
  • Cas Number : 522628-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205...207
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02009

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02009
Availability details for
Quantity
  • Availability
  • Add to Cart