ENA018105721 Enamine

4-Chloro-N-(3-chloro-quinoxalin-2-yl)-3-nitro-benzenesulfonamide

  • MDL number MFCD03651822
  • Cas Number : 522628-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02017

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02017
Availability details for
Quantity
  • Availability
  • Add to Cart