ENA018105733 Enamine

ethyl 5-(2-chloroacetamido)-4-cyano-3-methylthiophene-2-carboxylate

  • MDL number MFCD02965815
  • Cas Number : 449177-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02036
Availability details for
Quantity
  • Availability
  • Add to Cart