ENA018105734 Enamine

N-(3-Chloro-quinoxalin-2-yl)-4,N-dimethyl-benzenesulfonamide

  • MDL number MFCD00394772
  • Cas Number : 166271-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02037

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02037
Availability details for
Quantity
  • Availability
  • Add to Cart