ENA018105743 Enamine

ethyl 2-(2-chloroacetamido)-5-ethylthiophene-3-carboxylate

  • MDL number MFCD00276468
  • Cas Number : 219620-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02048
Availability details for
Quantity
  • Availability
  • Add to Cart