ENA018105755 Enamine

ethyl 5-(2-chloroacetyl)-2,4-dimethyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD01124420
  • Cas Number : 950-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02063
Availability details for
Quantity
  • Availability
  • Add to Cart