ENA018105759 Enamine

2-Chloro-N-[1-(2-chloro-phenyl)-2-oxo-cyclohexyl]-N-methyl-acetamide

  • MDL number MFCD03656358
  • Cas Number : 750611-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02068

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02068
Availability details for
Quantity
  • Availability
  • Add to Cart