ENA018105763 Enamine

2-chloro-1-[3-(2-chloroacetyl)-2,4,6-trimethylphenyl]ethan-1-one

  • MDL number MFCD06654903
  • Cas Number : 156641-43-9
  • Molecular Weight : 273.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02072
Availability details for
Quantity
  • Availability
  • Add to Cart