ENA018105771 Enamine

2-(2-Chloro-acetylamino)-5-methyl-4-p-tolyl-thiophene-3-carboxylic acid ethyl ester

  • MDL number MFCD03656351
  • Cas Number : 610274-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02081

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02081
Availability details for
Quantity
  • Availability
  • Add to Cart