ENA018105772 Enamine

2,4-dimethyl 5-(chlorosulfonyl)-3-methylthiophene-2,4-dicarboxylate

  • MDL number MFCD03656352
  • Cas Number : 750611-26-8
  • Molecular Weight : 312.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02082
Availability details for
Quantity
  • Availability
  • Add to Cart