ENA018105777 Enamine

Thiophene-2-sulfonic acid (2-amino-phenyl)-amide

  • MDL number MFCD02699247
  • Cas Number : 182499-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02087
Availability details for
Quantity
  • Availability
  • Add to Cart