ENA018105779 Enamine

3,4-diamino-N-(2-chlorophenyl)benzene-1-sulfonamide

  • MDL number MFCD02700611
  • Cas Number : 326023-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-191
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02089
Availability details for
Quantity
  • Availability
  • Add to Cart