ENA018105793 Enamine

3-(pyrrolidine-1-sulfonyl)aniline

  • MDL number MFCD02615287
  • Cas Number : 91619-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02111
Availability details for
Quantity
  • Availability
  • Add to Cart